Vill du logga in igen?
Beakers, low form
Leverantör: RSG Solingen
Stainless steel 18/10, with spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
ORDER
Expandera alla / Komprimera alla
Volym Höjd Ext.Ø Förp. VWR art.nr Antal
100 ml 65 mm 55 mm 1 213-1811
250 ml 80 mm 65 mm 1 RSGA070.250
500 ml 110 mm 80 mm 1 RSGA070.500
1000 ml 130 mm 100 mm 1 RSGA070.001
2000 ml 180 mm 120 mm 1 RSGA070.002
3000 ml 180 mm 150 mm 1 RSGA070.003
5000 ml 210 mm 180 mm 1 RSGA070.005
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal