Accessories
Leverantör: Pall Laboratory
ORDER
Expandera alla / Komprimera alla
Benämning Förp. VWR art.nr Antal
Fritted glass support base with No. 8 silicone stopper 1 516-7567
Fritted glass support base with sidearm 1 516-8922
Glas funnel, 300 ml for membrane filter funnel 47 mm Ø 1 516-7564
Glass flask, 1 l for membrane filter funnel 47 mm Ø 1 516-8923
Glass flask, 4 lt. for membrane filter funnel 47 mm Ø 1 516-7566
Glass funnel, 1 l for membrane filter funnel 47 mm Ø 1 516-7565
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal