Vill du logga in igen?
Capillary columns, VF-624ms
Leverantör: Agilent Technologies
VF-624ms columns set a new standard for the analysis of volatile organic compounds.
  • Improved phase technology reduces bleed, thereby increasing signal-to-noise ratios
  • Especially suited for analyzing solvents according to EPA methods 524, 624 and 8260, as well as USP 467
  • Eliminate ghost peaks and unstable baselines for best data accuracy
  • Enhanced selectivity eliminating co-eluters such as benzene and 1,2-dichloroethane for improved productivity
ORDER
Expandera alla / Komprimera alla
Column format Phase Skikttjocklek Innerdiam.×L Förp. VWR art.nr Antal
Capillary VF-624ms 0,84 µm 0,15 mm × 15 m 1 VARICP9101
Capillary VF-624ms 0,84 µm 0,15 mm × 20 m 1 VARICP9100
Capillary VF-624ms 1,40 µm 0,25 mm × 30 m 1 VARICP9102I5
Capillary VF-624ms 1,80 µm 0,32 mm × 30 m 1 VARICP9104
Capillary VF-624ms 1,80 µm 0,32 mm × 30 m 1 VARICP9104I5
Capillary VF-624ms 3,00 µm 0,53 mm × 30 m 1 VARICP9106
Capillary VF-624ms 3,00 µm 0,53 mm × 30 m 1 VARICP9106I5
Capillary VF-624ms 0,84 µm 0,15 mm × 30 m 1 VARICP9109
Capillary VF-624ms 0,84 µm 0,15 mm × 40 m 1 VARICP9110
Capillary VF-624ms 1,40 µm 0,25 mm × 60 m 1 VARICP9103
Capillary VF-624ms 1,40 µm 0,25 mm × 60 m 1 VARICP9103I5
Capillary VF-624ms 1,80 µm 0,32 mm × 60 m 1 VARICP9105
Capillary VF-624ms 3,00 µm 0,53 mm × 60 m 1 VARICP9107
Capillary VF-624ms 3,00 µm 0,53 mm × 75 m 1 VARICP9108
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal