Vill du logga in igen?
Capillary columns, CP-Molesieve 5Å
Leverantör: Agilent Technologies
This molecular-sieve coated capillary column is especially valuable when separating permanent gases.
  • Separate argon and oxygen at ambient temperature to reduce costs
  • High efficiency for increased productivity
  • Symmetrical peaks for accurate results
  • Analysis times are reduced by up to 75% compared to packed columns
  • High resolution analysis of permanent gases
ORDER
Expandera alla / Komprimera alla
Column format Phase Skikttjocklek Innerdiam.×L Förp. VWR art.nr Antal
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 10 m 1 VARICP6937
Capillary CP-Molesieve 5Å 30,00 µm 0,32 mm × 10 m 1 VARICP7535I5
Capillary CP-Molesieve 5Å 30,00 µm 0,32 mm × 10 m 1 VARICP7535
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 10 m 1 VARICP7537
Capillary CP-Molesieve 5Å 15,00 µm 0,53 mm × 15 m 1 VARICP7543
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 25 m 1 VARICP6938I5
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 25 m 1 VARICP6938
Capillary CP-Molesieve 5Å 30,00 µm 0,25 mm × 25 m 1 VARICP7533
Capillary CP-Molesieve 5Å 30,00 µm 0,32 mm × 25 m 1 VARICP7536
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 25 m 1 VARICP7538I5
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 25 m 1 VARICP7538
Capillary CP-Molesieve 5Å 10,00 µm 0,32 mm × 30 m 1 VARICP7534I5
Capillary CP-Molesieve 5Å 10,00 µm 0,32 mm × 30 m 1 VARICP7534
Capillary CP-Molesieve 5Å 15,00 µm 0,53 mm × 30 m 1 VARICP7544
Capillary CP-Molesieve 5Å 50,00 µm 0,53 mm × 50 m 1 VARICP7539
Capillary CP-Molesieve 5Å 30,00 µm 0,32 mm × 50 m 1 VARICP7540I5
Capillary CP-Molesieve 5Å 30,00 µm 0,32 mm × 50 m 1 VARICP7540
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal