Wall-mount mat roll dispensers, PIG®
Leverantör: New Pig
PIG®
NEWPMAT292EA 2000 SEK
NEWPMAT292
Wall-mount mat roll dispensers, PIG®
Dispenser för torkdukar
These mat roll dispensers feature a braking mechanism for more controlled dispensing.
  • Easy to refill and replace
  • Slot and cork brake prevents mat from unfurling as it's dispensed
  • Durable hot-rolled steel frame and galvanised core rod built to last in industrial settings