Respirator filters for masks, 7000 and 9000 Series, EasyLock®
Leverantör: Moldex
EasyLock®
111-1185EA 1275 SEK
111-1185 111-1184 111-1187 111-1186 111-1189 111-1190 111-1181 111-1183 111-1182 111-2305 111-0700 111-0699 111-0702 111-0701 111-0703
Respirator filters for masks, 7000 and 9000 Series, EasyLock®
Masker Andningsskydd, återanvändbara Maskfilter
EasyLock® filters protect against gas, vapour and dust. Suitable for series 7000 and 9000 masks.
  • Bayonet connector
  • Particulate filters can either be connected with the gas cartridges or directly with the mask body
  • Adaptors and pre-filters are not necessary
  • Simpler logistics - 30% less parts
  • Particulate filters with pleated filter technology: Significantly reduces breathing resistance, improves wearer comfort and reduces fatigue
Certifikat: EN 143:2000+A1:2006 (particulate filters)
EN 14387:2004+A1:2008 (gas filters and pre-assembled filters)