Vill du logga in igen?
Barium hexafluoroacetylacetonate
Leverantör: Alfa Aesar
Varning
14503.06EA 2350 SEK InStock
14503.06 14503.03
Barium hexafluoroacetylacetonate
Barium hexafluoroacetylacetonate
Formel: C₁₀H₂BaF₁₂O₄
MW: 551,44 g/mol
Smältpunkt: 220 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00075633
CAS nummer: 118131-57-0
EINECS: 000-000-0
UN: 1564
ADR: 6.1,III
Order Now
ORDER
SPECIFIKATIONER
RELATERAD INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal