Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) för syntes, Sigma-Aldrich®
Leverantör: Merck
M-Clarity™ quality level = MQ200
8.14991.0250EA 1600 SEK
8.14991.0250
Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) för syntes, Sigma-Aldrich®
Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers)
Formel: C₁₂H₁₄O₃
MW: 206,24 g/mol
MDL Number: MFCD00022338
CAS nummer: 77-83-8
EINECS: 201-061-8
Order Now
Specifikation testresultat
Melting Point 206.24 g/mol
Density 1.092 g/cm3 (20 °C)
Synonyms Ethyl 3-methyl-3-phenylglycidate
Hill Formula C12H14O3