Vill du logga in igen?
Potassium-4-(1-methyl-4-piperazinyl)phenyltrifluoroborate ≥95%
Leverantör: Alfa Aesar
Varning
H51884.06EA 7875 SEK InStock
H51884.06 H51884.03
Potassium-4-(1-methyl-4-piperazinyl)phenyltrifluoroborate ≥95%
Potassium-4-(1-methyl-4-piperazinyl)phenyltrifluoroborate
Formel: C₁₁H₁₅BF₃KN₂
MW: 282,16
Storage Temperature: Omgivande
MDL Number: MFCD16294522
EINECS: 000-000-0
UN: 0000
Order Now
ORDER
RELATERAD INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal