Vill du logga in igen?
Potassium-3-(furfurylaminocarbonyl)phenyltrifluoroborate
Leverantör: Alfa Aesar
Varning
H51182.03EA 810 SEK InStock
H51182.03 H51182.06
Potassium-3-(furfurylaminocarbonyl)phenyltrifluoroborate
Potassium-3-(furfurylaminocarbonyl)phenyltrifluoroborate
Formel: C₁₂H₁₀BF₃KNO₂
MW: 307,12
Storage Temperature: Omgivande
MDL Number: MFCD11977720
EINECS: 000-000-0
UN: 0000
Order Now
ORDER
RELATERAD INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal