2-Mercaptothiazole ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
TCIAM1739-1GEA 430 SEK
TCIAM1739-1G TCIAM1739-5G
2-Mercaptothiazole ≥98.0% (by GC, titration analysis)
2-Mercaptothiazole
Formel: C₃H₃NS₂
MW: 117,19 g/mol
MDL Number: MFCD00022449
CAS nummer: 82358-09-6
https://se.vwr.com/stibo/web/std.lang.all/60/75/28976075.jpg
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C