4'-Iodoacetanilide ≥95.0%
Leverantör: TCI EUROPE
Varning
TCIAI0846-1GEA 565 SEK
TCIAI0846-1G TCIAI0846-5G
4'-Iodoacetanilide ≥95.0%
4'-Iodoacetanilide
Formel: C₈H₈INO
MW: 261,0597 g/mol
MDL Number: MFCD00016352
CAS nummer: 622-50-4
https://se.vwr.com/stibo/web/std.lang.all/27/33/28972733.jpg
Order Now
Specifikation testresultat
Appearance White to Gray to Dark blue powder to crystal
Purity (GC) min. 95.0 %
Melting point 182.0 to 187.0 deg-C