Dimethylcyanamide 97%
Leverantör: Thermo Scientific
Fara
ACRO200210050EA 595 SEK
ACRO200210050 ACRO200211000
Dimethylcyanamide 97%
Dimethylcyanamide
Formel: C₃H₆N₂
Smältpunkt: –41 °C
Flampunkt: 58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001767
CAS nummer: 1467-79-4
EINECS: 215-991-7
UN: 2929
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥96.0 %