Dimethylcyanamide ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
TCIAD1150-25MLEA
2600
SEK
TCIAD1150-25ML
Dimethylcyanamide ≥98.0%
Dimethylcyanamide
Formel:
C₃H₆N₂ Kokpunkt: 164°C Flampunkt: 71°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00001767 CAS nummer: 1467-79-4 EINECS: 215-991-7 UN: 2810 ADR: 6.1,II |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...