Methyl thiosalicylate ≥98.0%
Leverantör: TCI EUROPE
Varning
TCIAM1881-25GEA 2800 SEK
TCIAM1881-25G TCIAM1881-5G
Methyl thiosalicylate ≥98.0%
Methyl thiosalicylate
Formel: C₈H₈O₂S
MW: 168,21 g/mol
Kokpunkt: 123°C
MDL Number: MFCD00060678
CAS nummer: 4892-02-8
https://se.vwr.com/stibo/web/std.lang.all/63/16/28976316.jpg
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %