2-Fluorophenylsulphur pentafluoride ≥97.0%
Leverantör: TCI EUROPE
Varning
TCIAF0823-1GEA 1875 SEK
TCIAF0823-1G
2-Fluorophenylsulphur pentafluoride ≥97.0%
2-Fluorophenylsulphur pentafluoride
Formel: C₆H₄F₆S
Kokpunkt: 94°C
Storage Temperature: Kylskåp
MDL Number: MFCD18909664
CAS nummer: 864230-02-4
UN: 1993
ADR: 3,III
https://se.vwr.com/stibo/web/std.lang.all/93/62/28969362.jpg
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %