Phenylsulphur pentafluoride ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
TCIAP1969-1GEA
1225
SEK
TCIAP1969-1G
Phenylsulphur pentafluoride ≥95.0%
Phenylsulphur pentafluoride
Formel:
C₆H₅F₅S Kokpunkt: 149°C Smältpunkt: -10°C Flampunkt: 48°C |
MDL Number:
MFCD09263740 CAS nummer: 2557-81-5 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...