Pinostilbene ≥97.0%
Leverantör: TCI EUROPE
TCIAP1927-100MGEA
2000
SEK
TCIAP1927-100MG
Pinostilbene ≥97.0%
Pinostilbene
Formel:
C₁₅H₁₄O₃ Storage Temperature: Kylskåp |
MDL Number:
MFCD12407153 CAS nummer: 42438-89-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...