2,4,5-Trichlorothioanisole ≥99.0%
Leverantör: TCI EUROPE
TCIAT0729-10GEA
1875
SEK
TCIAT0729-10G
2,4,5-Trichlorothioanisole ≥99.0%
2,4,5-Trichlorothioanisole
Formel:
C₇H₅Cl₃S |
MDL Number:
MFCD00044426 CAS nummer: 4163-78-4 EINECS: 224-009-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 60.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...