Vill du logga in igen?
Membrane filter dispenser, Sentino®
Leverantör: Pall Laboratory
Sentino™
Filter dispensers are ideal for high throughput microbiology testing laboratories looking for more ergonomic, faster ways to handle individual membranes. The Sentino® filter dispenser is designed to aseptically dispense one disc of membrane at the touch of a button.
  • Easy to load without jamming. Guide plates assist loading and keep refill web in alignment during use
  • Soft touch keypad for aseptically dispensing membrane discs
  • Designed to hold 200 membrane discs
  • Compact, portable design with miniaturised refill pack
  • Use plugged into main power source or charge battery for use anywhere. Lights indicate when battery charge is running low
  • Easy to clean
The Sentino® filter dispenser can be used for: Drinking and wastewater analysis, environmental and recreational water monitoring, personal care product manufacturing, beverage monitoring, pharmaceutical and industrial water system monitoring, pharmaceutical raw materials, intermediates and final product.
Leveransinformation: Delivery information: Supplied with power transformer, EU-plug, UK-plug and user guide CD.
Leveransinformation: Supplied with power transformer, EU-plug, UK-plug and user guide CD.
ORDER
Expandera alla / Komprimera alla
Benämning Förp. VWR art.nr Antal
Membrane filter dispenser, Sentino®, EU/UK-plug 1 516-0665Q
Dispenser pack refills
Typ Porstorlek Förp. VWR art.nr Antal
Supor® 0,2 µm 1.000 514-0629
GN-6 Metricel® 0,45 µm 1.000 514-0628
Metricel® Black 0,45 µm 1.000 514-0630
Metricel® Black 0,8 µm 1.000 514-0631
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Type Metricel® Black Supor® GN-6 Metricel®
Pore size 0,8 µm 0,45 µm 0,2 µm 0,45 µm
Thickness 135 µm 117 µm 145 µm 152 µm
Water flow rate ≥102,8 ml/min/cm² (0,7 bar) ≥34,5 ml/min/cm² (0,7 bar) ≥19,3 ml/min/cm² (0,7 bar) ≥65,0 ml/min/cm² (0,7 bar)
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal