Furfuryl methyl disulphide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
TCIAF0561-5GEA
685
SEK
TCIAF0561-5G
Furfuryl methyl disulphide ≥98.0%
Furfuryl methyl disulphide
Formel:
C₆H₈OS₂ MW: 160,2571 g/mol Kokpunkt: 61°C Flampunkt: 90°C |
MDL Number:
MFCD00012326 CAS nummer: 57500-00-2 EINECS: 260-773-7 |
Specifikation testresultat
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...