Cholesterol ethyl carbonate ≥94.0%
Leverantör: TCI EUROPE
TCIAC0716-10GEA
1725
SEK
TCIAC0716-10G
Cholesterol ethyl carbonate ≥94.0%
Cholesterol ethyl carbonate
Formel:
C₃₀H₅₀O₃ |
MDL Number:
MFCD0003770 CAS nummer: 23836-43-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 94.0 % |
Mesomorphic range | 78.0 to 83.0 deg-C |
Solubility in Chloroform | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...