Cholesterol heptyl carbonate

Leverantör: TCI EUROPE

TCIAB0264-10GEA 2750 SEK
TCIAB0264-10G
Cholesterol heptyl carbonate
Cholesterol heptyl carbonate
Formel: C₃₅H₆₀O₃
MDL Number: MFCD00059505
CAS nummer: 15455-81-9
https://se.vwr.com/stibo/web/std.lang.all/23/30/28932330.jpg

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Specific rotation [a]20/D -27.0 to -32.0 deg (C=2, CHCl3)
Solubility in Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR