4-Pentylphenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
TCIAA2336-5GEA
2050
SEK
TCIAA2336-5G
4-Pentylphenylboronic acid (contains varying amounts of Anhydride)
4-Pentylphenylboronic acid
Formel:
C₁₁H₁₇BO₂ MW: 192,07 g/mol |
MDL Number:
MFCD00995151 CAS nummer: 121219-12-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 95.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...