PIG® Poly Drip Decks
Leverantör: New Pig
PIG®
121-0187EA 6225 SEK
121-0187 121-0186 121-0185
PIG® Poly Drip Decks
Förvaring av tunnor
These poly drip decks capture drips from containers and drums before they cause slip-and-fall hazards. These decks keep leaky drums and containers from creating puddles and slick spots on the floors.
  • Low-profile 15 cm high design is easy to load or unload; pumps and funnels are easy to access
  • Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
  • Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
  • Translucent white sidewalls allow easy visual inspection
Partly made from post-industrial recycled polyethtylene.