Protein MW markers, Precise™
J383-200ULEA 1500 SEK
VWRCJ383-200UL
Protein MW markers, Precise™
Precise™ protein molecular weight marker offers seven bands ranging from 15,0 to 150,0 kDa. There is sufficient material for 40 assays.
  • 7 bands ranging from 15,0 to 150,0 kDa
  • Pre-mixed with loading buffer