Reactive Dye 5-packs, Amersham CyDye™
Leverantör: Cytiva
PA23501EA 9425 SEK
PA23501 PA23500 PA25501 PA25500 PA23001 PA25000 PA25001
Reactive Dye 5-packs, Amersham CyDye™
System för proteinlänkning och märkning
Mono-reactive dye pack contains derivatised CyDye™ that carries only one reactive group on each dye molecule for accurate labelling of amine groups, for example on antibodies or oligonucleotides.
  • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye™ fluorescent dyes
  • For use in ELISA, immunoprecipitation, and blotting applications
  • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labellings
Bis-reactive dye pack contains fluors carrying two reactive groups on each dye molecule for general labelling of protein amine groups.
Leveransinformation: Each kit contains five vials of dye. Each vial is sufficient for labelling 1 mg of protein or 30 nmol of amino groups.