Vill du logga in igen?
Pipette canisters, flat bottom
Leverantör: BELLCO GLASS INC.
Glass pipette canisters constructed of borosilicate glass.
  • With flat bottom
  • All components will withstand repeated dry or steam heat sterilization
  • Bottom silicone pad prevents chipping of pipette tips
ORDER
Expandera alla / Komprimera alla
Benämning Förp. VWR art.nr Antal
Pipette canister, with stainless steel closure, H×Ø: 430×38 mm 6 BELC1290-38430
Pipette canister, with stainless steel closure, recommended for Shortie pipettes, H×Ø: 250×38 mm 6 BELC1290-38250
Pipette canister (without closure), H×Ø: 300×38 mm 6 BELC1290-00300
Pipette canister (without closure), recommended for Shortie pipettes, H×Ø: 250×38 mm 6 BELC1290-00250
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
TILLBEHÖR
Expandera alla / Komprimera alla
Accessories
Benämning Förp. VWR art.nr Antal
Mounting plate, 90×90 mm, for use with 38mm OD glass pipette canisters 6 BELC1288-11111
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal