Flow cup viscosimeters, ASTM D-1200
Leverantör: Fungilab
645-0021EA 2950 SEK
645-0021 645-0023 645-0022
Flow cup viscosimeters, ASTM D-1200
Viskositetsmätare Viskosimetrar, utloppsbägartyp
Anodised aluminium cups with fixed stainless steel nozzles.
  • Rapid measurement and comparison of viscosities in the paint, ink and varnishing industries
  • Calibration certificates at specified temperatures included with the cups
  • Equations for relating flow times to viscosity
Designed for the measurement of kinematic viscosity by comparison of the break-thread flow times of similar liquids.
Certifikat: ASTM D1200
Beställningsinformation: Please order the stand and calibration fluids separately.