HPLC columns, Acclaim™ PepMap™ C8
Leverantör: Thermo Scientific
HYPE164723EA 6250 SEK
HYPE164723 HYPE164722 HYPE161185 HYPE164706 HYPE161194
HPLC columns, Acclaim™ PepMap™ C8
The Acclaim™ PepMap™ C8 reversed phase analytical columns are ideal for separation of very hydrophobic peptides like nontryptic peptides and are suitable for LC/MS applications.