Tubing connectors, T-shaped, Azlon®
Leverantör: SciLabware
AZLOCWH204EA 285 SEK
AZLOCWH204
Tubing connectors, T-shaped, Azlon®
Connectors for tubing, made of PP, T-shape.
  • Rigid tapered arms for firm grip
  • Good chemical resistance
  • Good temperature resistance