Special antifoam agent, deconex® 31 ANTIFOAM
Leverantör: Borer Chemie
DECO502600.00-F10WEA 0 SEK
DECO502600.00-F10W
Special antifoam agent, deconex® 31 ANTIFOAM
Special antifoam agent to prevent the development of foam. Suitable for automated and manual processes.
  • Prevents the development of foam caused by contaminations or residues such as proteins, fats and emulsifiers in water solutions containing wetting agents
  • Applicable in all pH ranges
  • High resistance to alkaline medium
  • EDTA-, NTA-, chlorine-, phosphate- and surfactant-free, no release of phosphates and surfactants into waste water
  • Technical support from the manufacturer and distributor