Handheld refractometers, OPTI+
Leverantör: Bellingham + Stanley
BELL38-H1EA 7425 SEK
BELL38-H1 BELL38-H2 BELL38-B8 BELL38-B9 BELL38-H3 BELL38-H4 BELL38-H5 BELL38-H6 BELL38-B1 BELL38-H7 BELL38-A1 BELL38-H8 BELL38-A2 BELL38-A3
Handheld refractometers, OPTI+
The OPTi+ refractometers cover the complete Brix range or equivalent scales.
  • Some models with ICUMSA (Brix)
  • Application specific temperature compensation (ATC)
  • Touch-of-a-button temperature display with °C or °F option
  • Unique measurement verification mode using long-life AG Fluids
  • Stainless steel prism dish for rapid sample temperature stabilisation
Certifikat: Calibration showing traceability to ICUMSA and NIST.
Leveransinformation: Supplied with multi-language user guide, certificate of calibration and protective case.