VWR®, Denhardt's solution, 50X, ultrapure
Leverantör: Avantor
VWR®
E717-50MLEA 0 SEK
E717-50ML
VWR®, Denhardt's solution, 50X, ultrapure
Buffertar Buffertar för elektrofores Hybridiseringsbuffert
A blocking reagent used to help prevent probe binding to open membrane space and non target nucleic acids.