First aid station
Leverantör: Orkla
118-0276EA 2150 SEK
118-0276
First aid station
Första hjälpen produkter Första Hjälpen-skåp
This first aid station has a transparent door with rubber edges, which protects against dust and dirt, thus making the equipment more hygienic and always ready for use. The station also features first aid instructions that are easy to follow. In addition, there are clear instructions on each product package.
  • Adhesive bandages (plasters) and wound cleansers are placed in dispensers for easy access and theft-proof storage
  • Plasters simply need to be pulled downwards, which avoids getting any dirt or blood on other areas
  • Transparent door makes it easy to see when a refill of any product is needed
  • Plaster refills and wound cleanser dispenser are locked in order to avoid unnecessary waste (key is included)
  • Space for additional products is provided behind a lid with first aid instructions
Contents:
2× Cederroth 4-in-1 Bloodstopper dressings
3× Cederroth 4-in-1 mini Bloodstopper dressings
1× Cederroth Protection Kit (aids in mouth-to-mouth resuscitation)
1× Salvequick plaster dispenser, including 45 sterile plastic plasters and 40 sterile fabric plasters
2× 20 Salvequick wound cleansers
1× 2 Burn gel dressings
1× Eye and wound-cleansing spray
First aid instructions
Refill key
Colour: Green.
W×D×H (mm): 290×120×560.