Vill du logga in igen?
Volumetric flasks, Class A, amber, white graduation
Leverantör: DURAN Group
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified.
  • White graduation, with one graduation mark
  • Certificate of conformity 'DE-M' mark
  • Available with individual certificate
*Vid hals
Logga in för att se avtalspriser och tillgänglighet.
  Kom ihåg mig i den här enheten
ORDER
Expandera alla / Komprimera alla
Volym NS Höjd Tolerans Förp. VWR art.nr Antal
5* ml 10/19 70 mm 0,04 ± ml 2 612-5332
10* ml 10/19 90 mm 0,04 ± ml 2 612-5337
20 ml 10/19 110 mm 0,04 ± ml 2 612-5338
25 ml 10/19 110 mm 0,04 ± ml 2 612-5339
50 ml 12/21 140 mm 0,06 ± ml 2 612-5340
100 ml 12/21 170 mm 0,1 ± ml 2 612-5352
100 ml 14/23 170 mm 0,1 ± ml 2 612-5353
200 ml 14/23 210 mm 0,15 ± ml 2 612-5354
250 ml 14/23 220 mm 0,15 ± ml 2 612-5355
500 ml 19/26 260 mm 0,25 ± ml 2 612-5356
1000 ml 24/29 300 mm 0,4 ± ml 1 612-5357
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal