Dialysis tubes equilibrium type, SP Bel-Art
Leverantör: Bel-Art Products
BELAH402610000EA 5825 SEK
BELAH402610000 BELAH402600000
Dialysis tubes equilibrium type, SP Bel-Art
Acrylic with stainless steel screws that tighten down on the dialysis membrane and also act as a gasket.
  • Filler hole screws on each half cavity are Teflon® PTFE coated stainless steel
  • Volume shown is for one half cell (double for total cell volume)
  • A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves