Vill du logga in igen?
Storage bags
Leverantör: hk-pack
LDPE transparent bags, ideal for storing and protecting food and small spare parts of all kinds.
  • Food safety is ensured
  • Flexible and tear resistant
  • Freeze-resistant starting 50 μ and heat resistant up to 80 °C
ORDER
Expandera alla / Komprimera alla
Benämning Mått Förp. VWR art.nr Antal
Food Storage bag 160×250 mm 1.000 HKPK100046
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal