VWR®, Carboys, Wide Neck
Leverantör: VWR
VWR®
216-0882EA 2400 SEK
216-0882 216-0820
VWR®, Carboys, Wide Neck
Flaskor Dunkar
HDPE, amber.
  • Compatible with interchangeable, leakproof VersaCap adapters
  • Features a wide mouth design for easy filling and cleaning
  • The carboy’s body includes design features that allow the user to easily hold and manipulate during lifting filling, and pouring
This amber carboys meet USP 671 requirements for light transmission, to allow for safe storage of photosensitive materials. HDPE carboys are made from a USP Class VI, FDA Grade, chemical resistant resin, making it ideal for most buffers and reagents as well as powders and solids.