Pocket safety cutter, disposable, SECUMAX POLYCUT
Leverantör: Martor
233-2226EA 52 SEK
MARO85002.08
Pocket safety cutter, disposable, SECUMAX POLYCUT
Steel blade, plastic handle.
  • Blade is safely embedded between the nose and the handle: Reduced risk for user
  • Disposable cutter: No need to change the blade
  • Suitable for left and right handers
  • Cutting depth: 3 mm
  • Non corrosive blade
This small, disposable cutter features an extra-long and pointed nose which enables comfortable and easy cutting of paper, foil, tape, cord. Also ideal for opening envelopes.