Anti-IFNAR1 Rabbit Polyclonal Antibody

Leverantör: Abgent
ABGEAP8550CEA 4050 SEK
ABGEAP8550C
Anti-IFNAR1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: IFNAR1 (interferon (alpha, beta and omega) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR