Anti-HNRNPA1 Rabbit Polyclonal Antibody

Leverantör: OriGene
ORIGTA314018EA 6550 SEK
ORIGTA314018
Anti-HNRNPA1 Rabbit Polyclonal Antibody
Antikroppar
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: HNRNPA1 (heterogeneous nuclear ribonucleoprotein A1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR