Anti-HTR2B Rabbit Polyclonal Antibody

Leverantör: OriGene
ORIGTA314042EA 6550 SEK
ORIGTA314042
Anti-HTR2B Rabbit Polyclonal Antibody
Antikroppar
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:00-1:500; ELISA: 1:1000

Type: Primary
Antigen: HTR2B (5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR