|
|
ORDER
SPECIFIKATIONER
Antigen symbol | ICAM2 |
Antigen name | intercellular adhesion molecule 2 |
Reaktivitet | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antikroppstyp | Primary |
Värd | Rabbit |
Isotype | IgG |
ELISA | Yes |
ImmunoChemistry | Yes |
Immunofluorescence | Yes |
Epitop | internal |
Cross adsorption | none |
Rening | purified by affinity chromatography |
Form | Liquid |
Storage buffer | Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol. |
Förvaringstemperatur | Cool |
Shipping temperature | Ice |