Anti-IGFBP2 Rabbit Polyclonal Antibody

Leverantör: OriGene
ORIGTA314059EA 6550 SEK
ORIGTA314059
Anti-IGFBP2 Rabbit Polyclonal Antibody
Antikroppar
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: IGFBP2 (insulin-like growth factor binding protein 2, 36kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR