Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-IGLL1 Rabbit Polyclonal Antibody

Leverantör: OriGene
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: IGLL1 (immunoglobulin lambda-like polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human


Logga in för att se avtalspriser och tillgänglighet.
  Kom ihåg mig i den här enheten
 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µl 100 µl ORIGTA314063
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol IGLL1
Antigen name immunoglobulin lambda-like polypeptide 1
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Antikroppstyp Primary
Värd Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Epitop internal
Cross Adsorption none
Rening purified by affinity chromatography
Form Liquid
Storage Buffer Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal