Anti-MXD4 Mouse Polyclonal Antibody
Leverantör: Abnova
ABNOH00010608-B02PEA 4575 SEK
ABNOH00010608-B02P
Anti-MXD4 Mouse Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: MXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human