Anti-STEAP4 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126393-200EA 0 SEK
BIRBORB126393-200 BIRBORB126393-100
Anti-STEAP4 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: STEAP4 (STEAP family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human