Anti-STON1 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126396-100EA 0 SEK
BIRBORB126396-100 BIRBORB126396-200
Anti-STON1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: STON1 (stonin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human