Anti-SYT16 Rabbit Polyclonal Antibody
Leverantör: Biorbyt
BIRBORB126398-100EA 0 SEK
BIRBORB126398-100 BIRBORB126398-200
Anti-SYT16 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: SYT16 (synaptotagmin XVI)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human